Learn More
J Kem Scientific Inc PROBE ADAPTER PTFE 1/8IN14/20
SDP
Catalog No. NC9430765
Encompass
Adapter, For between thermocouple probes and standard taper 14/20 joint, PTFE
Catalog No. NC9430765 Supplier J Kem Scientific Inc Supplier No. 1418
Adapter, For between thermocouple probes and standard taper 14/20 joint, PTFE