Learn More
Bellco Glass, Inc. Probe Adaptor,1.7
SDP
Catalog No. 501944939
Encompass_Preferred
316L stainless steel probe adapter extension, threaded. MADE IN THE USA. PG 13.5 THREAD X 1.7" EXTENSION
Catalog No. 50-194-4939 Supplier Bellco Glass, Inc. Supplier No. 196491005
316L stainless steel probe adapter extension, threaded. MADE IN THE USA. PG 13.5 THREAD X 1.7" EXTENSION