Learn More
Bellco Glass, Inc. Probe Adptr-old HP,1.5 Ext NPT Thread"
SDP
Catalog No. 501944938
Encompass_Preferred
316L stainless steel probe adapter extension, threaded. MADE IN THE USA.
Catalog No. 50-194-4938 Supplier Bellco Glass, Inc. Supplier No. 196491000
316L stainless steel probe adapter extension, threaded. MADE IN THE USA.