Learn More
Cole-Parmer Oakton ATC probe for pH 600, pH 610, pH 620, PC 650, PD 650, PCD 650 meters, 1-m
Catalog No. 501961315 Shop All Cole-Parmer Products
Encompass_Preferred
The probe provides ATC for most current Oakton pH meters. Corrosion-resistant stainless steel 30 K thermistor features durable polymer handle and 3.3 ft (1-m) cable. Probes measure 88-mm L x 3-mm dia; handle adds another 67 mm to the total length. Temperature range is 0 to 100C.
Catalog No. 50-196-1315 Supplier Cole-Parmer Supplier No. 3541805
The probe provides ATC for most current Oakton pH meters. Corrosion-resistant stainless steel 30 K thermistor features durable polymer handle and 3.3 ft (1-m) cable. Probes measure 88-mm L x 3-mm dia; handle adds another 67 mm to the total length. Temperature range is 0 to 100C.