Learn More
Roboz Surgical PROBE BLUNT SEPARATOR 6.25INP
SDP
Catalog No. NC9411702
Encompass
Probe & Blunt Separator; One End Semi-Blunt With Angled Tip, One End Tapered And Flat; 6-1/4 in. Length
Catalog No. NC9411702 Supplier Roboz Surgical Supplier No. RS9580
Probe & Blunt Separator; One End Semi-Blunt With Angled Tip, One End Tapered And Flat; 6-1/4 in. Length