Learn More
Leica Biosystems PROBE CLEANING KIT
Catalog No. NC0522148
Encompass_Preferred
The BOND Aspirating Probe Cleaning System contains reagents optimized to clean the aspirating probe of residual DAB.Sold in a standard reagent tray, the system is loaded onto BOND where a predefined cleaning protocol ensures maximum wash efficiency.
Catalog No. NC0522148 Supplier Leica Biosystems Supplier No. CS9100
The BOND Aspirating Probe Cleaning System contains reagents optimized to clean the aspirating probe of residual DAB.Sold in a standard reagent tray, the system is loaded onto BOND where a predefined cleaning protocol ensures maximum wash efficiency.