Learn More
Cole-Parmer Oakton Down-Well pH/CON/Temp Electrode with 10 ft Cable F/ PC10 Meter, 35 mm OD
Catalog No. 501961351 Shop All Cole-Parmer Products
Encompass_Preferred
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample. For your convenience, cables, power adapters, and other accessories are available for your Oakton meters and pocket testers.
Catalog No. 50-196-1351 Supplier Cole-Parmer Supplier No. 3563052
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample. For your convenience, cables, power adapters, and other accessories are available for your Oakton meters and pocket testers.