Learn More
Cole-Parmer Cole-Parmer Conductivity Probe, 4-cell, K = 1.0, Glass/Pt, 2.5 ft Cable
Catalog No. 501961238 Shop All Cole-Parmer Products
Encompass_Preferred
Keep your multiparameter meters up and running with replacement conductivity probes.
Catalog No. 50-196-1238 Supplier Cole-Parmer Supplier No. 3560892
Keep your multiparameter meters up and running with replacement conductivity probes.