Learn More
Glass Expansion Probe Connecting Line 0.18mm ID with EzyFit (Green/Black)
SDP
Catalog No. 502703018
Encompass_Preferred
Probe Connecting Line 0.18mm ID with EzyFit (Green/Black). Compatible with Glass Expansion Guardian Probe ASX-112FR (P/N 70-803-2029) for Teledyne Cetac ASX-112FR Autosampler. Creates a zero dead-volume connection between Guardian Probe and nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-900
Catalog No. 50-270-3018 Supplier Glass Expansion Supplier No. 708032085
Probe Connecting Line 0.18mm ID with EzyFit (Green/Black). Compatible with Glass Expansion Guardian Probe ASX-112FR (P/N 70-803-2029) for Teledyne Cetac ASX-112FR Autosampler. Creates a zero dead-volume connection between Guardian Probe and nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-900