Learn More
Glass Expansion Probe Connecting Line 0.25mm ID (Black)
SDP
Catalog No. 502703021
Encompass_Preferred
Probe Connecting Line 0.25mm ID (Black). For use with Glass Expansion Guardian Probe ASX-112FR (P/N 70-803-2029) and Teledyne Cetac ASX-112FR Autosampler. Creates a zero dead-volume connection between Guardian Probe and nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-900
Catalog No. 50-270-3021 Supplier Glass Expansion Supplier No. 708032030
Probe Connecting Line 0.25mm ID (Black). For use with Glass Expansion Guardian Probe ASX-112FR (P/N 70-803-2029) and Teledyne Cetac ASX-112FR Autosampler. Creates a zero dead-volume connection between Guardian Probe and nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-900