Learn More
Glass Expansion Probe Connecting Line 0.75mm ID (Red)
SDP
Catalog No. 502703038
Encompass_Preferred
Probe Connecting Line 0.75mm ID (Red). Compatible with Glass Expansion Guardian Probes for Aim Lab & Agilent SPS3/SPS4 Autosamplers (P/N 70-803-1957) and for Teledyne Cetac autosamplers (P/N 70-803-1803). Creates a zero dead-volume connection between Guardian Probe and nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-900
Catalog No. 50-270-3038 Supplier Glass Expansion Supplier No. 708031714
Probe Connecting Line 0.75mm ID (Red). Compatible with Glass Expansion Guardian Probes for Aim Lab & Agilent SPS3/SPS4 Autosamplers (P/N 70-803-1957) and for Teledyne Cetac autosamplers (P/N 70-803-1803). Creates a zero dead-volume connection between Guardian Probe and nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-900