Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp™ Probe Extension Cable
SDP

Catalog No. 11392014
Change view
Click to view available options
Length (Metric):
1 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-392-014 1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-392-014 Supplier Velp™ Supplier No. A00000002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Use With: Digital Thermoregulator VTF and VTF EVO

  • For use with VTF Digital Thermoregulator VTF
  • For use with VTF EVO Digital Thermoregulator

Specifications

For Use With (Equipment) Digital Thermoregulator VTF and VTF EVO
Length (Metric) 1 mm
Size 39.4 in.
Type Temperature Probe
Warranty 24 Months
Material Nickel-plated Zinc
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.