Learn More
Cole-Parmer Fischer Technology 604-175 Probe for Inside measurements, 0 to 40 mils, FGABI1.3-150, for models Deltascope FMP10 & Dualscope
Catalog No. NC1010655 Shop All Cole-Parmer Products
Encompass_Preferred
Accuracy: 0.5 m up to 50 m; < 1% above 50 m. Measuring method: Magnetic induction. Display: 240 x 160 pixels.
Catalog No. NC1010655 Supplier Cole-Parmer Supplier No. 5410007
Accuracy: 0.5 m up to 50 m; < 1% above 50 m. Measuring method: Magnetic induction. Display: 240 x 160 pixels.