Learn More
Perkin Elmer US LLC Probe for ST Nebulizers, 0.50 mm I.D.
Catalog No. 502104921
Encompass_Preferred
PTFE Sampling Probe Assembly with Carbon Fiber Support and Female Fitting, 0.50 mm ID for ICP-MS. Requires but does not include N8145017. Aspiration Rate: 400 µL/min Length: 100 cm
Catalog No. 50-210-4921 Supplier Perkin Elmer US LLC Supplier No. N8145140
PTFE Sampling Probe Assembly with Carbon Fiber Support and Female Fitting, 0.50 mm ID for ICP-MS. Requires but does not include N8145017. Aspiration Rate: 400 µL/min Length: 100 cm