Learn More
Roche Diagnostics KAPA PROBE FORCE qPCR Kits - 1000 x 20 uL reactions (Universal), 10 mL
Catalog No. 501965263 Shop All Roche Diagnostics Products
Encompass
KAPA PROBE FORCE qPCR Kit contains a highly inhibitor-resistant qPCR master mix that removes the need for DNA purification, enabling streamlined sample-to-Cq workflows. A third generation DNA polymerase engineered through our directed evolution technology imparts resistance to blood, tissue, and plant PCR inhibitors. This allows crude samples to be analyzed with comparable accuracy, reproducibility, and sensitivity as purified DNA. *This item is non returnable.
This item is not returnable. View return policy
Catalog No. 50-196-5263 Supplier Roche Diagnostics Supplier No. 07959346001
KAPA PROBE FORCE qPCR Kit contains a highly inhibitor-resistant qPCR master mix that removes the need for DNA purification, enabling streamlined sample-to-Cq workflows. A third generation DNA polymerase engineered through our directed evolution technology imparts resistance to blood, tissue, and plant PCR inhibitors. This allows crude samples to be analyzed with comparable accuracy, reproducibility, and sensitivity as purified DNA. *This item is non returnable.