Learn More
Waters Corp Probe Heater, APCI, IonSabre Probes, MS System
Catalog No. 502390505 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Probe Heater is a replacement APCI probe heater assembly designed for use with APCI and IonSabre APCI probes.
  • Replacement APCI probe heater assembly.
  • For use with APCI probes.
  • For use with IonSabre APCI probes.
  • Comes as a spare parts kit.
  • Includes 1 unit per package.
  • Mechanical type.
  • Designed for MS system type.
  • Replacement schedule is as required.
Catalog No. 50-239-0505 Supplier Waters Corp Supplier No. 700005687
The Probe Heater is a replacement APCI probe heater assembly designed for use with APCI and IonSabre APCI probes.
  • Replacement APCI probe heater assembly.
  • For use with APCI probes.
  • For use with IonSabre APCI probes.
  • Comes as a spare parts kit.
  • Includes 1 unit per package.
  • Mechanical type.
  • Designed for MS system type.
  • Replacement schedule is as required.