Learn More
Chemglass Life Sciences 12mm Probe Holder, Adjustable, M27 Thread
SDP
Catalog No. 501294453
Encompass
12mm Probe Holder, Adjustable, M27 Thread. Probe holders for 12mm probes. Height is adjustable with a compression fitting.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4453 Supplier Chemglass Life Sciences Supplier No. CLS138519
12mm Probe Holder, Adjustable, M27 Thread. Probe holders for 12mm probes. Height is adjustable with a compression fitting.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.