Learn More
Bellco Glass, Inc. Probe Holder,12mm OD For Vertical 45mm Sidearms
SDP
Catalog No. 501944780
Encompass_Preferred
45mm port fittingFor pg13.5 threaded probesFor vertical 45mm sidearmsMaterial: 316l SSTSupplied with hi-temperature s/cap, a523-608 & white silicone liner (a523-600). MADE IN THE USA
Catalog No. 50-194-4780 Supplier Bellco Glass, Inc. Supplier No. 196421580
45mm port fittingFor pg13.5 threaded probesFor vertical 45mm sidearmsMaterial: 316l SSTSupplied with hi-temperature s/cap, a523-608 & white silicone liner (a523-600). MADE IN THE USA