Learn More
Bellco Glass, Inc. Probe Holder,32mm
SDP
Catalog No. 501945130
Encompass_Preferred
•For use with Bellco BioProbe Spinner Flask Assemblies. MADE IN THE USA•Compression fitting accepts a 12mm OD probe•Monitoring of dissolved oxygen and pH can be achieved without interference from stirring paddles or port and thermowell assemblies•Includeso316L SST Compression Nuto316L SST Compression Bodyo12mm Teflon Ferruleo316L SST Hex Nuto316L SST Washero32mm White Silicone Washero32mm blue PP screw cap w/ 13/16 hole for compression fitting
Catalog No. 50-194-5130 Supplier Bellco Glass, Inc. Supplier No. 196597010
•For use with Bellco BioProbe Spinner Flask Assemblies. MADE IN THE USA•Compression fitting accepts a 12mm OD probe•Monitoring of dissolved oxygen and pH can be achieved without interference from stirring paddles or port and thermowell assemblies•Includeso316L SST Compression Nuto316L SST Compression Bodyo12mm Teflon Ferruleo316L SST Hex Nuto316L SST Washero32mm White Silicone Washero32mm blue PP screw cap w/ 13/16 hole for compression fitting