Learn More
Hach Company Electrode Holder for HQD Portable Meters (Standard electrodes only)
Catalog No. 501997184 Shop All Hach Company Products
Encompass
Designed for use in conjunction with the protective glove, electrode holders can be used to organise and store standard electrodes with 1 or 3 meter cables. Simply wrap cable around the holder and slide electrode in protective sheath. Protective glove can hold one or two electrode holders.
Catalog No. 50-199-7184 Supplier Hach Company Supplier No. 5829400
Designed for use in conjunction with the protective glove, electrode holders can be used to organise and store standard electrodes with 1 or 3 meter cables. Simply wrap cable around the holder and slide electrode in protective sheath. Protective glove can hold one or two electrode holders.