Learn More
Chemglass Life Sciences Probe Holder/Soaker Bottle
SDP
Catalog No. NC1420790
Encompass
The cap and gasket compresses and securely seals against the electrode body, sealing the solution from the atmosphere to minimize evaporation, prevent spillage and increase solution life. For use with 12mm diameter probes. Probe holder bottles should be filled with water for DO probe storage or KCI solution for pH electrode storage. Available in a pack of 4.
Catalog No. NC1420790 Supplier Chemglass Life Sciences Supplier No. CLS1435BTL
The cap and gasket compresses and securely seals against the electrode body, sealing the solution from the atmosphere to minimize evaporation, prevent spillage and increase solution life. For use with 12mm diameter probes. Probe holder bottles should be filled with water for DO probe storage or KCI solution for pH electrode storage. Available in a pack of 4.