Learn More
Ace Glass, Inc. Sensor probe, J thermocouple, 1/4in OD x 12in inconel sheath, 72in male mini plug lead
SDP
Catalog No. 50977453
Encompass
Sensor probe, J thermocouple, 1/4in OD x 12in inconel sheath, 72in male mini plug lead
Catalog No. 50-977-453 Supplier Ace Glass, Inc. Supplier No. 1211015
Sensor probe, J thermocouple, 1/4in OD x 12in inconel sheath, 72in male mini plug lead