Learn More
Cole-Parmer AEMC MN103 AC Clamp-On Current Probe, 1 mA-100 A, Lead
Supplier:  Cole-Parmer 2000205
Encompass
This clamp-on current proble ranges 100mA to 3000Amrs and measures AC, DC and AC/DC. It should be used with DMMs, oscilloscopes, dataloggers, handheld scopes and power meters. It has a voltage or current output.
Catalog No. 50-992-586