Learn More
EZKEM Probe PEEK 12" w/ Single Channel Tubing Assembly
SDP
Catalog No. NC1813034
Encompass_Preferred
EZkem’s 12 inch PEEK probe with pump and sampler tubing connections is generally used on CETAC and RA (AI Scientific) samplers for Alpkem and OI Analytical FSIII & FSIV auto analyzers, flow injection analysis, and segmented flow analysis. Tubing assembly measures 48 inches.
Catalog No. NC1813034 Supplier EZKEM Supplier No. A002536
EZkem’s 12 inch PEEK probe with pump and sampler tubing connections is generally used on CETAC and RA (AI Scientific) samplers for Alpkem and OI Analytical FSIII & FSIV auto analyzers, flow injection analysis, and segmented flow analysis. Tubing assembly measures 48 inches.