Learn More
Ace Glass, Inc. Sensor probe, Pt100 thermocouple, 1/4in OD x 12in Inconel sheath, 72in phone plug lead
SDP
Catalog No. 50977447
Encompass
Sensor probe, Pt100 thermocouple, 1/4in OD x 12in Inconel sheath, 72in phone plug lead
Catalog No. 50-977-447 Supplier Ace Glass, Inc. Supplier No. 1210318
Sensor probe, Pt100 thermocouple, 1/4in OD x 12in Inconel sheath, 72in phone plug lead