Learn More
Cole-Parmer Digi-Sense PROBE RTD 0.188)D W/PLUG
Supplier:  Cole-Parmer 3900975
Encompass_Preferred
These RTD assemblies consist of a single Low Range or High Range Wire Wound element constructed of 316 Stainless Steel sheath material, assembled together with an Aluminum screw cover connection head.
Catalog No. 50-196-1571