Learn More
Waters Corp Waters Probe Tip Assembly, 1 pk, Six-Month Replacement Schedule
Catalog No. 50792956 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This probe tip assembly is a replacement part designed for maintaining ESI probes in mass spectrometry systems. It includes the probe tip, gasket, and ferrule.
  • Replacement recommended every six months.
  • Includes probe tip, gasket, and ferrule.
  • One unit per package.
Catalog No. 50-792-956 Supplier Waters Corp Supplier No. 700004207
This probe tip assembly is a replacement part designed for maintaining ESI probes in mass spectrometry systems. It includes the probe tip, gasket, and ferrule.
  • Replacement recommended every six months.
  • Includes probe tip, gasket, and ferrule.
  • One unit per package.