Learn More
Cole-Parmer Digi-Sense Type-K, E x tra-Long SS Probe, Mini-Connector, 10" L. .125" Dia Tip, Grounded
Supplier:  Cole-Parmer 9375620
Encompass_Preferred
General-Purpose probes are designed for use with liquids, gases and semisolids. Used for a wide variety of applications and environments. Response Time: 2 seconds or greater. ABS handles provide a light weight design for ease of use along with durability. Probe handles feature a strain relief spring that protects the cable connection against damage due to repeated flexing and tugging.
Catalog No. NC9510755