Learn More
Cole-Parmer Digi-Sense TC/PR28875-3 TC/PR28875-3 Type K 90°-Angle Surface Thermocouple Probe with Guarded Tip
Supplier:  Cole-Parmer 0844510
Encompass_Preferred
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
Catalog No. 50-196-1493