Learn More
Sklar Instruments Probe with Eye, Sklar, 12 in., Stainless Steel. EA/1.
SDP
Catalog No. 501438148 Shop All Sklar Instruments Products
Encompass_Preferred
Probe with Eye, Sklar, 12 in., Stainless Steel. Sklar's Probe with Eye is a slender, slightly malleable, multipurpose instrument with a bulbous tip that can be used in a variety of procedures. It may be used to explore or gauge the depth of non-visible cavities, explore lesions, or manipulate tissue. This product is straight with an eye at the handle, and a length of 12 inches. The eye may be used for carrying ligatures.
This item is not returnable. View return policy
Catalog No. 50-143-8148 Supplier Sklar Instruments Supplier No. 302012
Probe with Eye, Sklar, 12 in., Stainless Steel. Sklar's Probe with Eye is a slender, slightly malleable, multipurpose instrument with a bulbous tip that can be used in a variety of procedures. It may be used to explore or gauge the depth of non-visible cavities, explore lesions, or manipulate tissue. This product is straight with an eye at the handle, and a length of 12 inches. The eye may be used for carrying ligatures.