Learn More
Med Vet International 6" Metal Dissection Probe, Blunt
Supplier:  Med Vet International PROBEB
Encompass_Preferred
Classic, thick, chrome plated, solid steel probe with hexagonal handle and angular blunt tip. Used for separating and tracking veins, nerves, and organs from connective tissue while dissecting.
Catalog No. 50-222-4031