Learn More
ATLANTIC BIOPHARM SOLUTIONS LLC PROBELESS DRY ICE MONITOR
Catalog No. NC3805855
Encompass_Preferred
Sensitech TT ULTRA DI AMB 2 DEC SU - MERCK CTS008070SINGLE USE, TempTale UltraProbeless Dry Ice Monitor, MERCK CTS008070 -95°C to +30°C (-139°F to +86°F)Low temp: -90 degrees CHigh temp : -69.5 degrees CMeasurement interval: 10 minutes,1.5 hour start up delay
Catalog No. NC3805855 Supplier ATLANTIC BIOPHARM SOLUTIONS LLC Supplier No. TUD2001071
Sensitech TT ULTRA DI AMB 2 DEC SU - MERCK CTS008070SINGLE USE, TempTale UltraProbeless Dry Ice Monitor, MERCK CTS008070 -95°C to +30°C (-139°F to +86°F)Low temp: -90 degrees CHigh temp : -69.5 degrees CMeasurement interval: 10 minutes,1.5 hour start up delay