Learn More
AdipoGen Probenazole
SDP
Catalog No. 502560236
Encompass_Preferred
Chemical. CAS 27605-76-1. Formula C10H9NO3S. MW 223.25. Synthetic. Benzothiazole fungicide with weak antibacterial activity. Active ingredient in Oryzemate, an agricultural chemical that has been widely used to protect rice plants from rice blast and the bacterial blight pathogen. Induces non-race specific resistance in rice plants against rice blast fungus. Defense activator that stimulates the SA/NPR1-mediated defense signaling pathway upstream of salicylic acid SA.
Catalog No. 50-256-0236 Supplier AdipoGen Supplier No. CDXP0128M100
Chemical. CAS 27605-76-1. Formula C10H9NO3S. MW 223.25. Synthetic. Benzothiazole fungicide with weak antibacterial activity. Active ingredient in Oryzemate, an agricultural chemical that has been widely used to protect rice plants from rice blast and the bacterial blight pathogen. Induces non-race specific resistance in rice plants against rice blast fungus. Defense activator that stimulates the SA/NPR1-mediated defense signaling pathway upstream of salicylic acid SA.