Learn More
Medchemexpress LLC Probenecid | 57-66-9 | 99.94% | 500 MG
SDP
Catalog No. 5000409137
Encompass_Preferred
Probenecid is a potent and selective agonist for transient receptor potential vanilloid 2 (TRPV2) channels and acts as an inhibitor of pannexin 1 channels. It is widely utilized in research to study various biological processes.
  • Potent and selective agonist of TRPV2 channels
  • Inhibits pannexin 1 channels
  • Efficiently inhibits ATP-dependent active vesicular NEM-GS uptake by MRP1 and MRP2
  • Inhibitor of hTAS2R16, hTAS2R38, and hTAS2R43 bitter taste receptors
  • Enhances cellular signals in GPCR calcium mobilization assays
  • Increases contractility (ejection fraction, EF) in WT mice
  • Involved in numerous clinical trials for various conditions
Catalog No. 50-004-09137 Supplier Medchemexpress LLC Supplier No. HYB0545500MG
Probenecid is a potent and selective agonist for transient receptor potential vanilloid 2 (TRPV2) channels and acts as an inhibitor of pannexin 1 channels. It is widely utilized in research to study various biological processes.
  • Potent and selective agonist of TRPV2 channels
  • Inhibits pannexin 1 channels
  • Efficiently inhibits ATP-dependent active vesicular NEM-GS uptake by MRP1 and MRP2
  • Inhibitor of hTAS2R16, hTAS2R38, and hTAS2R43 bitter taste receptors
  • Enhances cellular signals in GPCR calcium mobilization assays
  • Increases contractility (ejection fraction, EF) in WT mice
  • Involved in numerous clinical trials for various conditions