Learn More
Medchemexpress LLC Probenecid sodium | 23795-03-1 | MFCD01661360 | 99.4% | 307.34 | C13H18NNaO4S | 25 G
SDP
Catalog No. 5000281119
Encompass_Preferred
Probenecid sodium is the sodium salt of probenecid used in cell biology and pharmacology research as a selective TRPV2 agonist and an inhibitor of pannexin 1 channels. It is supplied as a high-purity research reagent with defined molecular properties and storage recommendations for stability.
  • Acts as a selective TRPV2 agonist and pannexin 1 inhibitor.
  • High purity (reported 99.4%).
  • Available in laboratory-scale package sizes, including 25 G.
  • Chemical formula C13H18NNaO4S; molecular weight 307.34 g/mol.
  • Recommended to store sealed and away from moisture; long-term stability in solvent at -80°C.
Catalog No. 50-002-81119 Supplier Medchemexpress LLC Supplier No. HYW40491625G
Probenecid sodium is the sodium salt of probenecid used in cell biology and pharmacology research as a selective TRPV2 agonist and an inhibitor of pannexin 1 channels. It is supplied as a high-purity research reagent with defined molecular properties and storage recommendations for stability.
  • Acts as a selective TRPV2 agonist and pannexin 1 inhibitor.
  • High purity (reported 99.4%).
  • Available in laboratory-scale package sizes, including 25 G.
  • Chemical formula C13H18NNaO4S; molecular weight 307.34 g/mol.
  • Recommended to store sealed and away from moisture; long-term stability in solvent at -80°C.