Learn More
Medchemexpress LLC Probenecid (sodium) | 23795-03-1 | MFCD01661360 | 99.4% | 307.34 g/mol | C13H18NNaO4S | 1 ML
SDP
Catalog No. 5000281806
Encompass_Preferred
Probenecid sodium is the sodium salt of probenecid supplied for research use. It is used as a TRPV2 agonist and in assays involving pannexin 1 channel inhibition. The product is offered as a solid or as a ready-to-use solution for laboratory applications.
  • Sodium salt form suitable for research applications.
  • Available as a 10 mM solution in DMSO (1 mL) or as a solid for reconstitution.
  • High purity (~99.4%).
  • Molecular formula C13H18NNaO4S, molecular weight 307.34 g/mol.
  • Acts as a TRPV2 agonist and inhibits pannexin 1 channels.
  • Supplied with safety data sheet and product documentation.
Catalog No. 50-002-81806 Supplier Medchemexpress LLC Supplier No. HYW40491610MM/1ML
Probenecid sodium is the sodium salt of probenecid supplied for research use. It is used as a TRPV2 agonist and in assays involving pannexin 1 channel inhibition. The product is offered as a solid or as a ready-to-use solution for laboratory applications.
  • Sodium salt form suitable for research applications.
  • Available as a 10 mM solution in DMSO (1 mL) or as a solid for reconstitution.
  • High purity (~99.4%).
  • Molecular formula C13H18NNaO4S, molecular weight 307.34 g/mol.
  • Acts as a TRPV2 agonist and inhibits pannexin 1 channels.
  • Supplied with safety data sheet and product documentation.