Learn More
Medchemexpress LLC Probenecid sodium | 23795-03-1 | MFCD01661360 | 99.4% | 307.34 g/mol | C13H18NNaO4S | 5 G
SDP
Supplier:  Medchemexpress LLC HYW4049165G
Encompass_Preferred
Probenecid sodium is the sodium salt of probenecid, a selective TRPV2 agonist that also inhibits pannexin 1 channels. Supplied as a white to off-white solid for research use only. Typical analytical purity is 99.4% and the compound is suitable for preparing concentrated DMSO stocks.
Catalog No. 50-002-81597