Learn More
Gold Biotechnology Inc ProBlock Bacter Protease Inhib
SDP
Catalog No. 5000562031
Encompass_Preferred
ProBlock Gold Bacterial Protease Inhibitor Cocktail contains optimized concentrations of various bacterial protease inhibitors which provide excellent inhibition of protease activities during protein purification from bacteria The cocktail contains both reversible and irreversible protease inhibitors to inhibit serine and cysteine proteases It also contains specific inhibitors for bacterial proteases such as aspartic proteases and aminopeptidases A 1X concentration of ProBlock Gold Bacterial inhibits over 90% of protease activities
Catalog No. 50-005-62031 Supplier Gold Biotechnology Inc Supplier No. GB3301
Add to Cart
Add to Cart
ProBlock Gold Bacterial Protease Inhibitor Cocktail contains optimized concentrations of various bacterial protease inhibitors which provide excellent inhibition of protease activities during protein purification from bacteria The cocktail contains both reversible and irreversible protease inhibitors to inhibit serine and cysteine proteases It also contains specific inhibitors for bacterial proteases such as aspartic proteases and aminopeptidases A 1X concentration of ProBlock Gold Bacterial inhibits over 90% of protease activities