Learn More
Gold Biotechnology Inc ProBlock Gold Protease Inhib
SDP
Supplier:  Gold Biotechnology Inc GB10810
Encompass_Preferred
Due to the optimized concentration of the various inhibitors the ProBlock Gold shows excellent inhibition of protease activities and is therefore suitable for the protection of proteins during purification from animal tissues plant tissues yeast and bacteria ProBlock Gold contains both irreversible and reversible protease inhibitors to inhibit serine cysteine and other proteases An separate bottle of EDTA is provided to inhibit metalloproteases
Catalog No. 50-005-62032