Learn More
Gold Biotechnology Inc ProBlock Gold Protease Inhib
SDP
Catalog No. 5000562116
Encompass_Preferred
Due to the optimized concentration of the various inhibitors the ProBlock Gold shows excellent inhibition of protease activities and is therefore suitable for the protection of proteins during purification from animal tissues plant tissues yeast and bacteria ProBlock Gold contains both irreversible and reversible protease inhibitors to inhibit serine cysteine and other proteases An separate bottle of EDTA is provided to inhibit metalloproteases
Catalog No. 50-005-62116 Supplier Gold Biotechnology Inc Supplier No. GB1082
Due to the optimized concentration of the various inhibitors the ProBlock Gold shows excellent inhibition of protease activities and is therefore suitable for the protection of proteins during purification from animal tissues plant tissues yeast and bacteria ProBlock Gold contains both irreversible and reversible protease inhibitors to inhibit serine cysteine and other proteases An separate bottle of EDTA is provided to inhibit metalloproteases