Learn More
Gold Biotechnology Inc ProBlock Protease Inhib EDTAfr
SDP
Catalog No. 5000562267
Encompass_Preferred
ProBlock -50 EDTA Free is designed for large volumes of extraction/lysis buffers and is supplied as 1 vial/kit (Cat GB-326-1) 10 vials /kit (Cat GB-326-10) or 20 vials/kit (Cat GB-326-20) with each vial suitable for 50 ml
Catalog No. 50-005-62267 Supplier Gold Biotechnology Inc Supplier No. GB32610
Add to Cart
Add to Cart
ProBlock -50 EDTA Free is designed for large volumes of extraction/lysis buffers and is supplied as 1 vial/kit (Cat GB-326-1) 10 vials /kit (Cat GB-326-10) or 20 vials/kit (Cat GB-326-20) with each vial suitable for 50 ml