Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human proBNP/NPPB ELISA Kit

Catalog No. EHPRONPPB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHPRONPPB 96 Tests
EHPRONPPBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EHPRONPPB Supplier Invitrogen™ Supplier No. EHPRONPPB
Add to Cart
Add to Cart

ELISA

The Human proBNP/NPPB ELISA quantitates Hu proBNP in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu proBNP. Principle of the method The Human proBNP solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

In cardiac tissue brain natriuretic peptide (BNP) is synthesized as 134 amino acid precursor (prepro-BNP), which is cleaved by proteases to form a 26 aa 'signal' peptide and a 108 aa pro-BNP. Proteolytic digestion of pro-BNP results in formation of 76 aa amino-terminal NT-proBNP and biologically active 32 aa BNP hormone molecule. Both proBNP and NTproBNP circulate in human plasma and have been proposed as markers for early diagnosis of left ventricular dysfunction as well as prognostic markers of possible cardiac complications at patients with heart failure.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P16860
Assay Range 0.137-100 ng/mL
Assay Sensitivity 0.14 ng/mL
Calibration Range 0.14 ng/mL to 100 ng/mL
Content And Storage Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody detects human proBNP and NT-proBNP. Other species not determined.
Sufficient For 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 5 kDa cardiac natriuretic peptide; AA408272; BNF; BNP; BNP(1-28); BNP(1-29); BNP(1-30); BNP(1-32); BNP(2-31); BNP(3-29); BNP(3-30); BNP(3-32); BNP(4-27); BNP(4-29); BNP(4-30); BNP(4-31); BNP(4-32); BNP(5-29); BNP(5-31); BNP(5-32); BNP32; BNP-34; Brain natriuretic factor; brain natriuretic peptide; Brain natriuretic peptide 32; Brain natriuretic peptide 45; brain type natriuretic peptide; gamma-brain natriuretic peptide; Iso-ANP; natriuretic peptide B; natriuretic peptide precursor B; natriuretic peptide precursor type B; natriuretic peptide type B; natriuretic peptides B; natriuretic protein; Nppb; NTproBNP; pro-BNP; type-B natriuretic factor5 kDa cardiac natriuretic peptide; AA408272; BNF; BNP; BNP(1-28); BNP(1-29); BNP(1-30); BNP(1-32); BNP(2-31); BNP(3-29); BNP(3-30); BNP(3-32); BNP(4-27); BNP(4-29); BNP(4-30); BNP(4-31); BNP(4-32); BNP(5-29); BNP(5-31); BNP(5-32); BNP32; BNP-34; Brain natriuretic factor; brain natriuretic peptide; Brain natriuretic peptide 32; Brain natriuretic peptide 45; brain type natriuretic peptide; gamma-brain natriuretic peptide; Iso-ANP; natriuretic peptide B; natriuretic peptide precursor B; natriuretic peptide precursor type B; natriuretic peptide type B; natriuretic peptides B; natriuretic protein; Nppb; NTproBNP; pro-BNP; type-B natriuretic factor; BNP
Gene ID (Entrez) 4879
Gene Symbol NPPB
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Gamma-brain natriuretic peptide, BNP(1-32), BNP-32
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Angiogenesis
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym BNP
Target pro-Brain Natriuretic Peptide
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.