Learn More
Selleck Chemical LLC Probucol-50mg
SDP
Catalog No. 502389799
Encompass_Preferred
Probucol (DH-581) is an anti-hyperlipidemic drug by lowering the level of cholesterol in the bloodstream by increasing the rate of LDL catabolism.
Catalog No. 50-238-9799 Supplier Selleck Chemical LLC Supplier No. S211950MG
Probucol (DH-581) is an anti-hyperlipidemic drug by lowering the level of cholesterol in the bloodstream by increasing the rate of LDL catabolism.