Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Probucol, 99.7%, MP Biomedicals™

Catalog No. ICN15637105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15637105 5 g
ICN15637150 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15637105 Supplier MP Biomedicals, Inc Supplier No. 0215637105
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23288-49-5
Molecular Formula C31H48O2S2
Molecular Weight (g/mol) 516.843
InChI Key FYPMFJGVHOHGLL-UHFFFAOYSA-N
Synonym probucol, lorelco, biphenabid, bisphenabid, lurselle, bisbid, lesterol, lursell, panavir, probucolum
PubChem CID 4912
ChEBI CHEBI:8427
IUPAC Name 2,6-ditert-butyl-4-[2-(3,5-ditert-butyl-4-hydroxyphenyl)sulfanylpropan-2-ylsulfanyl]phenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)SC(C)(C)SC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C

Specifications

Color White
Quantity 5 g
Formula Weight 516.8
Percent Purity 0.997
Physical Form Powder
Chemical Name or Material Probucol
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.