Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Probucol, MP Biomedicals™

Catalog No. MP215637110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215637110 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215637110 Supplier MP Biomedicals, Inc Supplier No. 0215637110
Add to Cart
Add to Cart

Enhances selective uptake of HDL-associated cholesteryl esters. Probucol, MP Biomedicals is an antioxidant, anti-inflammatory and hypocholesterolemic agent. It is also an antihyperlipoproteinemic .

Probucol inhibits atherogenesis in genetically hypercholesterolemic rabbits (Watanabe) and attenuates ischemia/reperfusion-induced cardiomyocyte apoptosis.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23288-49-5
Molecular Formula C31H48O2S2
Molecular Weight (g/mol) 516.843
InChI Key FYPMFJGVHOHGLL-UHFFFAOYSA-N
Synonym probucol, lorelco, biphenabid, bisphenabid, lurselle, bisbid, lesterol, lursell, panavir, probucolum
PubChem CID 4912
ChEBI CHEBI:8427
IUPAC Name 2,6-ditert-butyl-4-[2-(3,5-ditert-butyl-4-hydroxyphenyl)sulfanylpropan-2-ylsulfanyl]phenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)SC(C)(C)SC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C

Specifications

Melting Point 124°C to 126°C (literature)
Quantity 10 g
Solubility Information Soluble in ethanol (50mg/mL, clear and colorless solution).
Formula Weight 516.85
Percent Purity ≥98%
Physical Form Powder
Recommended Storage Room Temperature (15–30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.