Learn More
MilliporeSigma PROBUMIN DIAG GRD BSA 100G
Supplier:  MilliporeSigma 820451
Encompass
Probumin Bovine Serum Albumin Diagnostic Grade, Powder, size 100 g. Manufactured by a proprietary heat-shock fractionation process, this BSA powder is subjected to an additional heat-shock to control for proteolytic activity.
Catalog No. NC0582624