Learn More
Aviva Systems Biology Corp PROC Antibody - N-terminal region (ARP41701_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41701_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PROC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PROC. Alternate names: PROC1, PC, APCApplicationWB. Species reactivity: Cow, Horse, Human, Rat
Catalog No. 50-014-57960