Learn More
Siemens Healthineers ProC™ Control
Catalog No. 23044677 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
6 x 1 mL
Catalog No. Quantity
23-044-677 6 x 1 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-677 Supplier Siemens Healthineers Supplier No. 10446097
Add to Cart
Add to Cart
This item is not returnable. View return policy
Assayed control to monitor the performance of Factor V Leiden assay in the pathological range.
ProC™ Control Plasma is an assayed control used to monitor the performance of the Factor V Leiden assay in the pathological range.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Control
For Use With (Application) Coagulation Testing
Quantity 6 x 1 mL