Learn More
YMC America, Inc ProC18 col 120A 5um 300x4.6mm
SDP
Catalog No. 5000880065
Encompass_Preferred
YMC-Pack Pro C18 columns start with a tightly controlled highly purified silica gel followed by a high-coverage C18 bonding and finished with a unique endcapping procedure utilizing Lewis acid/Lewis base chemistry Together these features provide unequaled separations with sharp and symmetrical peaks for all types of organic molecules especially basic pharmaceuticals and chelating compounds
Catalog No. 50-008-80065 Supplier YMC America, Inc Supplier No. AS12S053046WT
YMC-Pack Pro C18 columns start with a tightly controlled highly purified silica gel followed by a high-coverage C18 bonding and finished with a unique endcapping procedure utilizing Lewis acid/Lewis base chemistry Together these features provide unequaled separations with sharp and symmetrical peaks for all types of organic molecules especially basic pharmaceuticals and chelating compounds