Learn More
YMC America, Inc ProC18 guard 120A 10um 10x4mm
SDP
Catalog No. 5000880066
Encompass
YMC-Pack Pro C18 columns start with a tightly controlled highly purified silica gel followed by a high-coverage C18 bonding and finished with a unique endcapping procedure utilizing Lewis acid/Lewis base chemistry Together these features provide unequaled separations with sharp and symmetrical peaks for all types of organic molecules especially basic pharmaceuticals and chelating compounds
Catalog No. 50-008-80066 Supplier YMC America, Inc Supplier No. AS12S110104GC
YMC-Pack Pro C18 columns start with a tightly controlled highly purified silica gel followed by a high-coverage C18 bonding and finished with a unique endcapping procedure utilizing Lewis acid/Lewis base chemistry Together these features provide unequaled separations with sharp and symmetrical peaks for all types of organic molecules especially basic pharmaceuticals and chelating compounds